missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium dihydrogentrifluoride, 50-55% w/w soln. in 1,2-dichloroethane

Product Code. 11354815
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11354815 5 g 5g
11344815 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11354815 Supplier Thermo Scientific Alfa Aesar Supplier No. L17891.06
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 99337-56-1 | C16H38F3N | 301.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 99337-56-1
Molecular Formula C16H38F3N
Molecular Weight (g/mol) 301.48
MDL Number MFCD00145365
InChI Key MRXQMNWIADOAJY-UHFFFAOYSA-M
Synonym tetra-n-butylammonium dihydrogen trifluoride, tetra-n-butylammonium dihydrogentrifluoride, dihydrogen tetrabutylammonium fluoride, tetrabutylammonium dihydrogentrifluoride, tetrabutylammonium dihydrogen trifluoride, tetrabutylammonium fluoride dihydrofluoride, tetrabutylazanium fluoride dihydrofluoride, tetrabutylammoniumdihydrogentrifluoride, tetrabutylamonium dihydrogentrifluoride, tetrabutylammoniumdihydrogen trifluoride
PubChem CID 11748636
SMILES F.F.[F-].CCCC[N+](CCCC)(CCCC)CCCC

Tekniske data

Flash Point 13°C (55°F)
Quantity 5 g
UN Number UN2924
IUPAC Name tetrabutylazanium dihydrofluoride fluoride
Formula Weight 301.48
Concentration or Composition (by Analyte or Components) 50 to 55% w/w soln. in 1,2-dichloroethane
Chemical Name or Material Tetra-n-butylammonium dihydrogentrifluoride
compliance-icons compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • Tetra-n-butylammonium dihydrogentrifluoride, 50-55% w/w soln. in 1,2-dichloroethane
Signalord
  • Danger
Farekategori
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 C
  • Specific target organ toxicity Category 3
Faresætning
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H350-May cause cancer.
Sikkerhedssætning
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplerende oplysninger
  • MIXTURE LIST-Contain: 1,2-Dichloroethane,Tetra-n-butylammonium dihydrogentrifluoride

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.