missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium hydroxide, 55% w/w aq. soln.

Product Code. 11337158
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11337158 50 g 50g
11347158 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11337158 Supplier Thermo Scientific Alfa Aesar Supplier No. 016198.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

As a reagent for ion-pair extractions | CAS: 2052-49-5 | C16H37NO | 259.48 g/mol

As a reagent for ion-pair extractionsTetra-n-butylammonium hydroxide, 55% w/w aq. solution is used as a phase-transfer catalyst in organic synthesis especially for alkylation and condensation reactions. It is used as a curing accelerator for epoxy resins. It is also used in preparation of dichlorocarbene from chloroform.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2052-49-5
Molecular Formula C16H37NO
Molecular Weight (g/mol) 259.48
MDL Number MFCD00009425
InChI Key VDZOOKBUILJEDG-UHFFFAOYSA-M
Synonym tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
PubChem CID 2723671
SMILES [OH-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Color Colorless to Yellow
Density 0.97 g/mL
pH 14
Flash Point None
Linear Formula [CH3(CH2)3]4NOH
Refractive Index 1.435
Quantity 50 g
UN Number UN3267
Beilstein 3571228
Sensitivity Air Sensitive
Solubility Information Miscible with water and organic solvents.
IUPAC Name tetrabutylazanium;hydroxide
Formula Weight 259.48
Concentration or Composition (by Analyte or Components) 55% w/w aq. soln.
Physical Form Liquid
Chemical Name or Material Tetra-n-butylammonium hydroxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.