missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium iodide, 98%

Product Code. 11492857
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11492857 50 g 50g
11402867 250 g 250g
11482857 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11492857 Supplier Thermo Scientific Alfa Aesar Supplier No. A15484.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 311-28-4 | C16H36IN | 369.38 g/mol

Tetra-n-butylammonium iodide is used as a catalyst for methylation and benzylation of alcohols with dimethyl sulfate and benzyl bromide respectively. It is a useful source of iodide anion and involved in the preparation of neopentyl iodides from the triflates. It is also used in the preparation of novel quaternary amines, which serve as an antibacterial agent in the rise of drug-resistant bacteria. It acts as a phosphonium reversible inhibitors of cholinesterases. It is associated with boron trifluoride diethyl¬ etherate and used in the cleavage of ethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetra-n-butylammonium iodide is used as a catalyst for methylation and benzylation of alcohols with dimethyl sulfate and benzyl bromide respectively. It is a useful source of iodide anion and involved in the preparation of neopentyl iodides from the triflates. It is also used in the preparation of novel quaternary amines, which serve as an antibacterial agent in the rise of drug-resistant bacteria. It acts as a phosphonium reversible inhibitors of cholinesterases. It is associated with boron trifluoride diethyl¬ etherate and used in the cleavage of ethers.

Solubility
Soluble in water and methanol. Insoluble in benzene.

Notes
Light sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 311-28-4
Molecular Formula C16H36IN
Molecular Weight (g/mol) 369.38
MDL Number MFCD00011636
InChI Key DPKBAXPHAYBPRL-UHFFFAOYSA-M
Synonym tetrabutylammonium iodide, tetra-n-butylammonium iodide, tbai, tetrabutylazanium iodide, tetra-n-butylammoniumjodid, tetrabutyl ammonium iodide, 1-butanaminium, n,n,n-tributyl-, iodide, n,n,n-tributyl-1-butanaminium iodide, tetra-n-butylammoniumjodid czech, tetrabutylammonium ion iodide
PubChem CID 67553
IUPAC Name tetrabutylazanium;iodide
SMILES [I-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 142°C to 148°C
Quantity 50 g
Beilstein 3916152
Sensitivity Hygroscopic; Light sensitive
Solubility Information Soluble in water and methanol. Insoluble in benzene.
Formula Weight 369.38
Percent Purity 98%
Chemical Name or Material Tetra-n-butylammonium iodide
compliance-icons
Product Identifier
  • Tetra-n-butylammonium iodide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Tetrabutylammonium iodide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.