missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium phosphate, 98%, non-UV

Product Code. 15442757
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15442757 2 g 2g
15452757 10 g 10g
15462757 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15442757 Supplier Thermo Scientific Alfa Aesar Supplier No. 046746.04
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5574-97-0 | C16H38NO4P | 339.46 g/mol

Tetra-n-butylammonium phosphate, is used in chemicals, pharmaceutical research, pharmaceutical intermediates, and reagents. Tetrabutylammonium phosphate can be used in organic synthesis. It is used as an Ion-associating reagent for HPLC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5574-97-0
Molecular Formula C16H38NO4P
Molecular Weight (g/mol) 339.46
MDL Number MFCD00064526
InChI Key ARRNBPCNZJXHRJ-UHFFFAOYSA-M
Synonym tetrabutylammonium dihydrogen phosphate, tetrabutylammonium phosphate, tetrabutylammonium dihydrogenphosphate, 1-butanaminium, n,n,n-tributyl-, phosphate 1:1, tetra-n-butylammonium phosphate, tetrabutylazanium dihydrogen phosphate, dihydrogen phosphate; tetrabutylazanium, dihydrogen phosphate; tetrabutylammonium, tetrabutyl ammonium dihydrogen phosphate, tetrabutylammonium ion dihydrogenphosphate ion
PubChem CID 2735142
IUPAC Name dihydrogen phosphate;tetrabutylazanium
SMILES OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 151°C to 154°C
Color White
Linear Formula [CH3(CH2)3]4NH2PO4
Quantity 2 g
Beilstein 5196532
Solubility Information It is soluble in water.
Formula Weight 339.46
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Tetra-n-butylammonium phosphate non-UV

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.