missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium tetrafluoroborate, 99%

Product Code. 15413147
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15413147 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15413147 Supplier Thermo Scientific Alfa Aesar Supplier No. A16688.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 429-42-5 | C16H36BF4N | 329.27 g/mol

As Pharmaceutical Intermediates. Also in catalysis. Reasonably effiicient (1500-2000 plates/m), long-llved open tubular columns were prepared by using the organic molten salt tetra-n-butylammonlum tetrafluoroborate as the stationary phase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
As Pharmaceutical Intermediates. Also in catalysis. Reasonably effiicient (1500-2000 plates/m), long-llved open tubular columns were prepared by using the organic molten salt tetra-n-butylammonlum tetrafluoroborate as the stationary phase.

Solubility
Slightly soluble in Water, methanol( 0.1 g/mL), ethanol (100 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Specifications

CAS 429-42-5
Melting Point 159°C to 162°C
Odor Odorless
Molecular Formula C16H36BF4N
Linear Formula [CH3(CH2)3]4NBF4
MDL Number MFCD00011634
Quantity 100 g
Beilstein 3577508
Synonym tetrabutylammonium tetrafluoroborate, tetra-n-butylammonium tetrafluoroborate, tetrabutylammonium fluoroborate, 1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-, tetrabutyl ammonium tetrafluoroborate
Solubility Information Slightly soluble aq. soln.,methanol(0.1 g/mL),ethanol (100 mg/ml).
InChI Key NNZZSJSQYOFZAM-UHFFFAOYSA-N
SMILES F[B-](F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC
IUPAC Name tetrabutylazanium; tetrafluoroboranuide
Molecular Weight (g/mol) 329.27
PubChem CID 67932
Formula Weight 329.27
Percent Purity 99%
Chemical Name or Material Tetra-n-butylammonium tetrafluoroborate
Show More Show Less
compliance-icons
Product Identifier
  • Tetra-n-butylammonium tetrafluoroborate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 1-Butanaminium, N,N,N-tributyl-tetrafluoroborate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.