missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium trifluoromethanesulfonate, 99%

Product Code. 11494483
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11494483

Brand: Thermo Scientific Alfa Aesar A11013.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 35895-70-6 | C18H36F3NO2 | 355.49 g/mol

Used as a catalyst for the reactions of condensation of alcohols and carboxylic acids, reaction of aromatic compounds with sulfonyl chlorides, cracking of alkanes, alkylation of alkenes, isomerisation of alkanes and trans-alkylation of aromatics, trans-bromination and other Friedel-Crafts reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a catalyst for the reactions of condensation of alcohols and carboxylic acids, reaction of aromatic compounds with sulfonyl chlorides, cracking of alkanes, alkylation of alkenes, isomerisation of alkanes and trans-alkylation of aromatics, trans-bromination and other Friedel-Crafts reactions.

Solubility
Miscible in watetr. Soluble in methanol(0.1 g/mL).

Notes
Hygroscopic. Keep container tightly closed in a cool, dry and well-ventilated place. Store away from oxidizing agents, air and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35895-70-6
Molecular Formula C18H36F3NO2
Molecular Weight (g/mol) 355.49
MDL Number MFCD00042585
InChI Key WTEXQPWIUJQYJQ-UHFFFAOYSA-M
Synonym tetrabutylammonium triflate, tetrabutylammonium trifluoromethanesulfonate, tetra-n-butylammonium trifluoromethanesulfonate, tetrabutylammonium ion triflate, trifluoromethanesulfonic acid tetrabutylammonium salt, acmc-1aglb, tetrakis but-1-yl ammonium triflate, tetrabutylazanium; trifluoromethanesulfonate, tetra-n-butylammonium trifluoromethanesulphonate
PubChem CID 2733306
IUPAC Name tetrabutylazanium;trifluoromethanesulfonate
SMILES [O-]C(=O)C(F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 112°C to 115°C
Quantity 25 g
Beilstein 4172978
Sensitivity Hygroscopic
Solubility Information Miscible in watetr. Soluble in methanol(0.1g/mL).
Formula Weight 391.54
Percent Purity 99%
Chemical Name or Material Tetra-n-butylammonium trifluoromethanesulfonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.