missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylphosphonium hydroxide, 40% w/w aq. soln.

Product Code. 11348868
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
Unit Size:
1 L
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348868 50 mL 50mL
11358868 250 mL 250mL
11338868 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11348868 Supplier Thermo Scientific Alfa Aesar Supplier No. 041718.AD
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 14518-69-5 | C16H37OP | 276.45 g/mol

Tetra-n-butylphosphonium hydroxide is used for the extraction of wheat straw and dissolution of wood. It is also used to produce hydroxide ion or proton conductive ionic liquids by mixing it with benzimidazole. It can be used as a phase transfer catalyst in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14518-69-5
Molecular Formula C16H37OP
Molecular Weight (g/mol) 276.45
MDL Number MFCD00068456
InChI Key DFQPZDGUFQJANM-UHFFFAOYSA-M
Synonym tetrabutylphosphonium hydroxide, tbph, tetrabutylphosphonium hydroxide solution, phosphonium,tetrabutyl-, hydroxide 1:1, phosphonium, tetrabutyl-, hydroxide 1:1, acmc-1c8ln, tetrabutylphosphoniumhydroxide, tetrabutylphosphanium hydroxide, tetra-n-butylphosphonium hydroxide w/w aq. soln., tetrabutylphosphonium hydroxide solution, 40 wt. % in h2o
PubChem CID 84487
IUPAC Name tetrabutylphosphanium;hydroxide
SMILES [OH-].CCCC[P+](CCCC)(CCCC)CCCC

Specifications

Color Colorless to Yellow
Density 0.989 g/mL
Linear Formula [CH3(CH2)3]4POH
Refractive Index 1.412
Quantity 50 mL
UN Number UN3267
Solubility Information Miscible with water.
Formula Weight 276.45
Concentration or Composition (by Analyte or Components) 40% w/w aq. soln.
Physical Form Liquid
Chemical Name or Material Tetra-n-butylphosphonium hydroxide
Show More Show Less
compliance-icons
Product Identifier
  • Tetra-n-butylphosphonium hydroxide, 40% w/w aqueous solution
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Tetrabutylphosphonium hydroxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.