missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-heptylammonium iodide, 99%, Thermo Scientific™

Product Code. 11499220
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499220 100 g 100g
11479220 5 g 5g
11489220 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499220 Supplier Thermo Scientific Alfa Aesar Supplier No. A16642.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3535-83-9
Molecular Formula C28H60IN
Molecular Weight (g/mol) 537.699
MDL Number MFCD00041984
InChI Key KCSOHLKZTZMKQA-UHFFFAOYSA-M
Synonym tetraheptylammonium iodide, tetra-n-heptylammonium iodide, 1-heptanaminium, n,n,n-triheptyl-, iodide, tetraheptylazanium iodide, 1-heptanaminium, n,n,n-triheptyl-, iodide 1:1, ammonium, tetraheptyl-, iodide, acmc-1aeuw, 1-heptanaminium,n,n-triheptyl-, iodide, tetraheptylammonium iodide at
PubChem CID 77071
IUPAC Name tetraheptylazanium;iodide
SMILES CCCCCCC[N+](CCCCCCC)(CCCCCCC)CCCCCCC.[I-]

Specifications

Melting Point 122°C to 124°C
Quantity 100 g
Beilstein 4167115
Sensitivity Hygroscopic; Light sensitive
Formula Weight 537.7
Percent Purity 99%
Chemical Name or Material Tetra-n-heptylammonium iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.