missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-hexylammonium iodide, 99%

Product Code. 11355109
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11355109 25 g 25g
11345109 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11355109 Supplier Thermo Scientific Alfa Aesar Supplier No. A17589.14
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2138-24-1 | C24H52IN | 481.591 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2138-24-1
Molecular Formula C24H52IN
Molecular Weight (g/mol) 481.591
MDL Number MFCD00041981
InChI Key VRKHAMWCGMJAMI-UHFFFAOYSA-M
Synonym tetrahexylammonium iodide, tetra-n-hexylammonium iodide, 1-hexanaminium, n,n,n-trihexyl-, iodide, tetrahexylazanium iodide, 1-hexanaminium, n,n,n-trihexyl-, iodide 1:1, acmc-209fjk, tetrahexyl ammonium iodide, tetra-n-hexyl ammonium iodide, ammonium, tetrahexyl-, iodide, 1-hexanaminium,n,n-trihexyl-, iodide
PubChem CID 75056
IUPAC Name tetrahexylazanium;iodide
SMILES CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC.[I-]

Specifications

Melting Point 99°C to 105°C
Quantity 25 g
Sensitivity Hygroscopic; Light sensitive
Formula Weight 481.59
Percent Purity 99%
Chemical Name or Material Tetra-n-hexylammonium iodide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.