missing translation for 'onlineSavingsMsg'
Learn More

Tetra-N-octylammonium Iodide, TRC

Product Code. 30009976
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009976 100 mg 100mg
30009557 500 mg 500mg
30008487 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009976 Supplier Toronto Research Chemicals Supplier No. TRCT305888100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16829-91-7
Molecular Formula C32 H68 N . I
Molecular Weight (g/mol) 593.793
Synonym N,N,N-trioctyl-1-Octanaminium iodide (1:1), N,N,N-trioctyl-1-Octanaminium iodide (9CI), tetraoctyl-Ammonium iodide (8CI), Tetraoctylammonium iodide (6CI)
IUPAC Name tetraoctylazanium;iodide
SMILES [I-].CCCCCCCC[N+](CCCCCCCC)(CCCCCCCC)CCCCCCCC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C32H68N.HI/c1-5-9-13-17-21-25-29-33(30-26-22-18-14-10-6-2,31-27-23-19-15-11-7-3)32-28-24-20-16-12-8-4;/h5-32H2,1-4H3;1H/q+1;/p-1
Formula Weight 593.44
Chemical Name or Material Tetra-N-octylammonium Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.