missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-propylammonium hydroxide, 1M aq. soln.

Product Code. 11303476
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11303476 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11303476 Supplier Thermo Scientific Alfa Aesar Supplier No. L14328.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4499-86-9 | C12H29NO | 203.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4499-86-9
Density 1
Molecular Formula C12H29NO
MDL Number MFCD00009360
Quantity 500 g
UN Number UN3267
Beilstein 4207983
Sensitivity Air Sensitive
Synonym tetrapropylammonium hydroxide, tetra-n-propylammonium hydroxide, 1-propanaminium, n,n,n-tripropyl-, hydroxide, tpaoh, tetrapropylammonium oxide
InChI Key LPSKDVINWQNWFE-UHFFFAOYSA-M
SMILES [OH-].CCC[N+](CCC)(CCC)CCC
IUPAC Name tetrapropylazanium hydroxide
Molecular Weight (g/mol) 203.37
PubChem CID 20586
Formula Weight 203.37
Concentration or Composition (by Analyte or Components) 1 M aq. soln.
Chemical Name or Material Tetra-n-propylammonium hydroxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.