missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-propylammonium hydroxide, 40% w/w aq. soln.

Product Code. 11303357
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11303357 25 g 25g
11323357 5 g 5g
11333357 100 g 100g
11313357 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11303357 Supplier Thermo Scientific Alfa Aesar Supplier No. 017456.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4499-86-9 | C12H29NO | 203.37 g/mol

Tetra-n-propylammonium hydroxide is used as an intermediate, ion exchange agent, surface-active agent and solvent. Further, it is used as an antistatic agent, detergent sanitizers, softener for textiles and paper products, emulsifying agents and pigment dispersers. It finds application as curing accelerators. In addition to this, it acts as a phase transfer catalyst in organic synthesis to enhance the reaction rate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4499-86-9
Molecular Formula C12H29NO
Molecular Weight (g/mol) 203.37
MDL Number MFCD00009360
InChI Key LPSKDVINWQNWFE-UHFFFAOYSA-M
Synonym tetrapropylammonium hydroxide, tetra-n-propylammonium hydroxide, 1-propanaminium, n,n,n-tripropyl-, hydroxide, tpaoh, tetrapropylammonium oxide, unii-t686luy7nk, tetrapropylazanium hydroxide, t686luy7nk, ammonium, tetrapropyl-, hydroxide, n,n,n-tripropyl-1-propanaminium hydroxide
PubChem CID 20586
IUPAC Name tetrapropylazanium;hydroxide
SMILES [OH-].CCC[N+](CCC)(CCC)CCC

Specifications

Color Colorless to Yellow
Density 0.99 g/mL
Boiling Point 102°C
Linear Formula (CH3CH2CH2)4NOH
Refractive Index 1.3716
Quantity 25 g
UN Number UN3267
Beilstein 4207983
Sensitivity Air Sensitive
Solubility Information Miscible with strong electrolytes.
Formula Weight 203.37
Concentration or Composition (by Analyte or Components) 40% w/w aq. soln.
Physical Form Liquid
Chemical Name or Material Tetra-n-propylammonium hydroxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.