missing translation for 'onlineSavingsMsg'
Learn More

Tetrabenzyl pyrophosphate, 98%

Product Code. 11397216
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397216 1 g 1g
11307226 250 mg 250mg
15498889 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11397216 Supplier Thermo Scientific Alfa Aesar Supplier No. L09083.03
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 990-91-0 | C28H28O7P2 | 538.473 g/mol

Tetrabenzyl pyrophosphate is used in the preparation of phosphoryl derivatives of shikimic acid in the presence of LDA. It is used in the preparation of dibenzyl phosphoro fluoridate in the presence of cesium fluoride as a catalyst. It is also used as a precursor in pharmaceuticals and involved in the phosphorylation of inositol derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrabenzyl pyrophosphate is used in the preparation of phosphoryl derivatives of shikimic acid in the presence of LDA. It is used in the preparation of dibenzyl phosphoro fluoridate in the presence of cesium fluoride as a catalyst. It is also used as a precursor in pharmaceuticals and involved in the phosphorylation of inositol derivatives.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acid and bases and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 990-91-0
Molecular Formula C28H28O7P2
Molecular Weight (g/mol) 538.473
MDL Number MFCD00051941
InChI Key NSBNXCZCLRBQTA-UHFFFAOYSA-N
Synonym tetrabenzyl pyrophosphate, tetrabenzyl diphosphate, pyrophosphoric acid tetrabenzyl ester, tetrabenzylpyrophosphate, diphosphoric acid, tetrakis phenylmethyl ester, dibenzyl dibenzyloxyphosphoryl oxyphosphonate, benzyl pyrophosphate, acmc-209sbp, tetra benzyl pyrophosphate, tetrabenzyl diphosphate #
PubChem CID 563183
IUPAC Name dibenzyl bis(phenylmethoxy)phosphoryl phosphate
SMILES C1=CC=C(C=C1)COP(=O)(OCC2=CC=CC=C2)OP(=O)(OCC3=CC=CC=C3)OCC4=CC=CC=C4

Specifications

Melting Point 58°C to 64°C
Quantity 1 g
UN Number UN3261
Beilstein 2068292
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 538.48
Percent Purity 98%
Chemical Name or Material Tetrabenzyl pyrophosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.