missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tetrabromophenol Blue

Product Code. 11430651
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430651 1 g 1g
11440651 5 g 5g
11450651 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11430651 Supplier Thermo Scientific Chemicals Supplier No. B20123.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4430-25-5 | C19H6Br8O5S | 985.544 g/mol

Tetrabromophenol Blue is used as acid-base indicator. It is used in oxidative- as well as in non-oxidative hair dye formulations. It is used as an industrial dye, a laboratory indicator, and a biological stain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrabromophenol Blue is used as acid-base indicator. It is used in oxidative- as well as in non-oxidative hair dye formulations. It is used as an industrial dye, a laboratory indicator, and a biological stain.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4430-25-5
Molecular Formula C19H6Br8O5S
Molecular Weight (g/mol) 985.544
MDL Number MFCD00005876
InChI Key QPMIVFWZGPTDPN-UHFFFAOYSA-N
Synonym 3,4,5,6,3',5',3'',5''-Octabromophenolsulfonphthalein
PubChem CID 78159
IUPAC Name 2,6-dibromo-4-[4,5,6,7-tetrabromo-3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
SMILES C1=C(C=C(C(=C1Br)O)Br)C2(C3=C(C(=C(C(=C3Br)Br)Br)Br)S(=O)(=O)O2)C4=CC(=C(C(=C4)Br)O)Br

Specifications

Melting Point 203°C to 205°C (decomposition)
Beilstein 378438
Solubility Information Soluble in water.
Formula Weight 985.55
Quantity 1 g
Chemical Name or Material Tetrabromophenol Blue

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.