missing translation for 'onlineSavingsMsg'
Learn More

Tetrabromophenol Blue, TRC

Product Code. 30134569
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134569 100 mg 100mg
30134582 50 mg 50mg
30009501 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134569 Supplier Toronto Research Chemicals Supplier No. TRCT295638100MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4430-25-5
Molecular Formula C19 H6 Br8 O5 S
Molecular Weight (g/mol) 985.54
Synonym Phenol, 4,4'-(4,5,6,7-tetrabromo-3H-2,1-benzoxathiol-3-ylidene)bis[2,6-dibromo-, S,S-dioxide, Tetrabromophenol blue (6CI), 3H-2,1-Benzoxathiole, phenol deriv., 4,4’-(4,5,6,7-Tetrabromo-1,1-dioxido-3H-2,1-benzoxathiol-3-ylidene)bis[2,6-dibromophenol], NSC 11236
IUPAC Name 2,6-dibromo-4-[4,5,6,7-tetrabromo-3-(3,5-dibromo-4-hydroxy-phenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
SMILES Oc1c(Br)cc(cc1Br)C2(OS(=O)(=O)c3c(Br)c(Br)c(Br)c(Br)c23)c4cc(Br)c(O)c(Br)c4

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C19H6Br8O5S/c20-7-1-5(2-8(21)16(7)28)19(6-3-9(22)17(29)10(23)4-6)11-12(24)13(25)14(26)15(27)18(11)33(30,31)32-19/h1-4,28-29H
Formula Weight 977.3403
Chemical Name or Material Tetrabromophenol Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.