missing translation for 'onlineSavingsMsg'
Learn More

Tetrabromophenolphthalein ethyl ester, ACS reagent, Thermo Scientific™

Product Code. 11935141 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11935141 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11935141 Supplier Thermo Scientific Acros Supplier No. 445890010
Quantity Price
1 144.00 € / 1g
10 132.21 € / 1g

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1176-74-5
Melting Point 208.0°C to 211.0°C
Molecular Formula C22H13Br4KO4
Quantity 1 g
Merck Index 15, 9331
Solubility Information Solubility aq. soln.: negligible.
InChI Key WCIQBKUTYDIBJC-UHFFFAOYSA-M
SMILES [K+].CCOC(=O)C1=CC=CC=C1C(C1=CC(Br)=C([O-])C(Br)=C1)=C1C=C(Br)C(=O)C(Br)=C1
IUPAC Name potassium 2,6-dibromo-4-[(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)[2-(ethoxycarbonyl)phenyl]methyl]benzen-1-olate
Molecular Weight (g/mol) 700.06
Formula Weight 661.96
Grade ACS Reagent
Chemical Name or Material Tetrabromophenolphthalein ethyl ester
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.