missing translation for 'onlineSavingsMsg'
Learn More

Tetrabromothiophene, 99%

Product Code. 11449913
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449913 25 g 25g
11439913 5 g 5g
11459913 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11449913 Supplier Thermo Scientific Alfa Aesar Supplier No. A13009.14
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 3958-03-0 | C4Br4S | 399.72 g/mol

Tetrabromothiophene, holds wide applications in organic electronics and photonics. It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrabromothiophene, holds wide applications in organic electronics and photonics. It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water. Soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong bases, strong oxidizing agents. Stable under normal temperatures and pressures.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3958-03-0
Molecular Formula C4Br4S
Molecular Weight (g/mol) 399.72
MDL Number MFCD00005419
InChI Key AVPWUAFYDNQGNZ-UHFFFAOYSA-N
Synonym tetrabromothiophene, perbromothiophene, thiophene, tetrabromo, tetrabromthiophen, 2,3,4,5-tetrabromo-thiophene, pubchem7661, acmc-1ae1j, 2,4,5-tetrabromothiophene, ksc491i3p, 2,3,4,5-tetrabromothiophen
PubChem CID 77565
IUPAC Name 2,3,4,5-tetrabromothiophene
SMILES C1(=C(SC(=C1Br)Br)Br)Br

Specifications

Melting Point 113°C to 117°C
Boiling Point 324°C to 327°C
Odor Odorless
Quantity 25 g
Beilstein 383664
Solubility Information Insoluble in water. Soluble in organic solvents.
Formula Weight 399.72
Percent Purity 99%
Chemical Name or Material Tetrabromothiophene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.