missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutyl orthosilicate, 95%, ACROS Organics™

Product Code. 10153351 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10153351 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10153351 Supplier Thermo Scientific Acros Supplier No. 313690050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 4766-57-8
Color Colorless
Density 0.8990g/mL
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C16H36O4Si
Linear Formula Si(OC4H9)4
Refractive Index 1.4115 to 1.4145 (20°C, 589nm)
MDL Number MFCD00009432
Quantity 5 g
Synonym tetrabutyl orthosilicate, tetrabutoxysilane, tetra-n-butoxysilane, silane, tetrabutoxy, silicon tetrabutoxide, butyl silicate buo 4si, silicic acid h4sio4 , tetrabutyl ester, butyl silicate c4h9o 4si, butyl silicate, tetrabutyl orthosilice
InChI Key UQMOLLPKNHFRAC-UHFFFAOYSA-N
SMILES CCCCO[Si](OCCCC)(OCCCC)OCCCC
IUPAC Name tetrabutyl silicate
Molecular Weight (g/mol) 320.545
PubChem CID 78500
Formula Weight 320.54
Percent Purity ≥94%
Physical Form Liquid
Chemical Name or Material Tetrabutyl orthosilicate, 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.