missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium bromide, 99+%

Product Code. 10646202 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
2.5kg
25g
500g
50g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10646202 25 g Glass bottle 25g
10586783 50 g Glass bottle 50g
10327940 100 g Glass bottle 100g
10266000 500 g Glass bottle 500g
11345932 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10646202 Supplier Thermo Scientific Acros Supplier No. 185680250
  • ONLINE EXCLUSIVE PRICE
    30.90 €
    29.00 € / 25g
    Save 1.90 €
    6% Off
View all promotions
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1643-19-2 | C16H36BrN | 322.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1643-19-2
Molecular Formula C16H36BrN
Molecular Weight (g/mol) 322.36
MDL Number MFCD00011633
InChI Key JRMUNVKIHCOMHV-UHFFFAOYSA-M
Synonym tetrabutylammonium bromide, tetra-n-butylammonium bromide, tbab, tetrabutylazanium bromide, tetrabutyl ammonium bromide, tetrabutyl-ammonium bromide, aliquat 100, tbabr, ipc-tba-br, unii-vjz168i98r
PubChem CID 74236
ChEBI CHEBI:51993
IUPAC Name tetrabutylazanium;bromide
SMILES CCCC[N+](CCCC)(CCCC)CCCC.[Br-]

Specifications

Melting Point 100.0°C to 104.0°C
Color Cream to White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (Argentometry)
Packaging Glass bottle
Linear Formula [CH3(CH2)3]4NBr
Quantity 25 g
Fieser 04,477; 05,644; 07,353; 17,363
Solubility Information Solubility in water: 600g/L (20°C).
Formula Weight 322.36
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium bromide, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • Tetrabutylammonium bromide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Tetrabutylammonium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.