missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium chloride hydrate, 96%

Product Code. 15189293 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
5g
Product Code. Quantity Packaging unitSize
15189293 100 g Glass Bottle 100g
10704092 5 g Glass Bottle 5g
10714092 25 g Glass Bottle 25g
3 options
This item is not returnable. View return policy

Product Code. 15189293

Brand: Thermo Scientific Acros 432061000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Phase transfer catalyst | CAS: 37451-68-6 | C16H36ClN | 277.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37451-68-6
Molecular Formula C16H36ClN
Molecular Weight (g/mol) 277.92
MDL Number MFCD00011635
InChI Key NHGXDBSUJJNIRV-UHFFFAOYSA-M
Synonym tetrabutylammonium chloride hydrate, tetrabutyl ammonium chloride hydrate, tetrabutylammonium chloride monohydrate, tetra-n-butylammonium chloride hydrate, tetrabutylammoniumchlorideh2o, 1-butanaminium, n,n,n-tributyl-, chloride, monohydrate, tbac hydrate, bu4ncl.h2o, n-bu4ncl.h2o, ksc222g0b
PubChem CID 16212273
IUPAC Name tetrabutylazanium;chloride;hydrate
SMILES [Cl-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 37.0°C to 39.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95% min. (On dried substance) (Argentometry)
Packaging Glass Bottle
Quantity 100 g
Formula Weight 295.94
Percent Purity 96%
Physical Form Low Melting Solid
Chemical Name or Material Tetrabutylammonium Chloride Hydrate
compliance-icons
Product Identifier
  • Tetrabutylammonium chloride hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.