missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium fluoride trihydrate, 99%

Product Code. p-195721 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10022970 10 g Glass bottle 10g
10266320 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10022970 Supplier Thermo Scientific Acros Supplier No. 221080100
Add to basket
This item is not returnable. View return policy

CAS: 87749-50-6 | C16H36FN·3H2O | 315.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87749-50-6
Molecular Formula C16H36FN·3H2O
Molecular Weight (g/mol) 315.51
MDL Number MFCD00149981
InChI Key VEPTXBCIDSFGBF-UHFFFAOYSA-M
Synonym tetrabutylammonium fluoride trihydrate, tetra-n-butylammonium fluoride trihydrate, tetrabutylazanium fluoride trihydrate, tetrabutylammonium trihydrate fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride, trihydrate, tetrabutylammonium ion trihydrate fluoride, tetra-n-butylammonium fluoride hydrate, bu4nf trihydrate, pubchem2275, pubchem4057
PubChem CID 11726816
IUPAC Name tetrabutylazanium;fluoride;trihydrate
SMILES CCCC[N+](CCCC)(CCCC)CCCC.O.O.O.[F-]

Specifications

Melting Point 58°C to 60°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula [CH3(CH2)3]4NF·3H2O
Quantity 10 g
Fieser 04,477; 05,645; 07,353; 08,467; 09,444; 10,378; 11,499; 12,458; 14,293; 15,168; 17,321
Solubility Information Solubility in water: soluble
Formula Weight 315.51
Percent Purity 99%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material Tetrabutylammonium fluoride trihydrate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Tetrabutylammonium fluoride trihydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.