missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Hydrogen Sulfate, 0.98, HPLC Grade, J.T. Baker™

Product Code. 15598904 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Wide Mouth Amber Glass Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15598904 500 g Wide Mouth Amber Glass Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15598904 Supplier Avantor J.T.Baker Supplier No. V36007

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High purity solvents for use in analytical and preparative separations

    For Use in Ion-Pair Chromatography, High Performance Liquid Chromatography; For Laboratory, Research, or Manufacturing Use

Specifications

CAS 32503-27-8
Packaging Wide Mouth Amber Glass Bottle
Molecular Formula C16H37NO4S
Quantity 500 g
Specific Gravity 1L = 0.00kg
Synonym tetrabutylammonium hydrogen sulfate, tetrabutylammonium hydrogensulfate, tetrabutylammonium bisulfate, tetra-n-butylammonium hydrogen sulfate, tbahs, tetrabutylammonium sulfate 1:1, 1-butanaminium, n,n,n-tributyl-, sulfate 1:1, tetrabutylammonium hydrogen sulphate, tetrabutylazanium hydrogen sulfate, ipc-tba-hs
InChI Key SHFJWMWCIHQNCP-UHFFFAOYSA-M
SMILES OS([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC
IUPAC Name hydrogen sulfate;tetrabutylazanium
Molecular Weight (g/mol) 339.54
PubChem CID 94433
Formula Weight 339.54
Percent Purity 98%
Grade HPLC
Chemical Name or Material Tetrabutylammonium Hydrogen Sulfate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.