missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Hydrogensulfate, TRC

Product Code. 30426579
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30426579 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30426579 Supplier Toronto Research Chemicals Supplier No. TRCT28922050G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetrabutylammonium Hydrogensulfate
CAS 32503-27-8
Purity Grade Notes HPLC
Molecular Formula C16 H36 N . H O4 S
Quantity 50 g
Synonym N,N,N-Tributyl-1-butanaminium Sulfate, Tetrabutyl-ammonium Sulfate, Tetrabutylammonium Hydrogen Sulfate, TBAHS-MB, Tetra-n-butylammonium Hydrogen Sulfate, Tetrabutylammonium Bisulfate, Tetrabutylammonium Hydrosulfate
InChI Formula InChI=1S/C16H36N.H2O4S/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-5(2,3)4/h5-16H2,1-4H3;(H2,1,2,3,4)/q+1;/p-1
SMILES CCCC[N+](CCCC)(CCCC)CCCC.OS(=O)(=O)[O-]
IUPAC Name hydrogen sulfate;tetrabutylazanium
Molecular Weight (g/mol) 339.5343
Formula Weight 339.2443
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.