missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabutylammonium hydroxide, 1M solution in methanol, AcroSeal™
Shop All Thermo Scientific Chemicals ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 65.0°C |
| Chemical Name or Material | Tetrabutylammonium hydroxide |
| Melting Point | -98.0°C |
| CAS | 2052-49-5, 67-56-1 |
| Color | Colorless to Yellow |
| Physical Form | Solution |
| Quantity | 100 mL |
| Flash Point | 12°C |
| Packaging | AcroSeal™ Glass Bottle |
| Refractive Index | 1.3775 |
| Show More |
Safety and Handling
Product Identifier
- Tetrabutylammonium hydroxide, 1M solution in methanol
- Danger
- Acute toxicity Category 3
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity after single exposure Category 1
- H370-Causes damage to organs.
- H314-Causes severe skin burns and eye damage.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?