missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium hydroxide, 40 wt.% solution in methanol

Product Code. 10339323 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10339323 2.5 L Glass bottle 2.5 L
10130182 100 mL Glass bottle 100mL
10242902 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10339323 Supplier Thermo Scientific Acros Supplier No. 366320025
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2052-49-5 | C16H37NO | 259.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetrabutylammonium hydroxide
Name Note 40 wt.% solution in methanol
CAS 2052-49-5, 67-56-1
Melting Point -98.0°C
Color Colorless to Yellow
Density 0.9100g/mL
Boiling Point 65.0°C
Flash Point 12°C
Packaging Glass bottle
Molecular Formula C16H37NO
Linear Formula [CH3(CH2)3]4NOH
MDL Number MFCD00009425
Quantity 2.5 L
Beilstein 04, II, 634
Fieser 05,645; 11,500
Specific Gravity 0.91
Synonym tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
Solubility Information Solubility in water: soluble
InChI Key VDZOOKBUILJEDG-UHFFFAOYSA-M
SMILES [OH-].CCCC[N+](CCCC)(CCCC)CCCC
IUPAC Name tetrabutylazanium;hydroxide
Molecular Weight (g/mol) 259.48
PubChem CID 2723671
Formula Weight 259.46
Percent Purity ≥40%
Physical Form Solution
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetrabutylammonium hydroxide, 40 wt.% solution in methanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H370-Causes damage to organs.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.