missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium iodide, 98%

Product Code. 10498700 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
2.5kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10498700 25 g Glass bottle 25g
10225830 100 g Glass bottle 100g
10773541 500 g Glass bottle 500g
11985441 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10498700 Supplier Thermo Scientific Acros Supplier No. 161270250
  • ONLINE EXCLUSIVE PRICE
    24.15 €
    22.60 € / 25g
    Save 1.55 €
    6% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 311-28-4 | C16H36IN | 369.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 311-28-4
Molecular Formula C16H36IN
Molecular Weight (g/mol) 369.38
MDL Number MFCD00011636
InChI Key DPKBAXPHAYBPRL-UHFFFAOYSA-M
Synonym tetrabutylammonium iodide, tetra-n-butylammonium iodide, tbai, tetrabutylazanium iodide, tetra-n-butylammoniumjodid, tetrabutyl ammonium iodide, 1-butanaminium, n,n,n-tributyl-, iodide, n,n,n-tributyl-1-butanaminium iodide, tetra-n-butylammoniumjodid czech, tetrabutylammonium ion iodide
PubChem CID 67553
IUPAC Name tetrabutylazanium;iodide
SMILES [I-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 141°C to 150°C
Color White to Cream
Ignition Residue 1% max.
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula [CH3(CH2)3]4NI
Quantity 25 g
Fieser 05,646; 06,566; 07,355; 09,447; 11,501
Solubility Information Solubility in water: <5%
Formula Weight 369.36
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium iodide
Show More Show Less
compliance-icons
Product Identifier
  • Tetrabutylammonium iodide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.