missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Phosphate, HPLC Grade, J.T. Baker™

Product Code. 15578834 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Wide Mouth Amber Glass Bottle
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15578834 25 g Wide Mouth Amber Glass Bottle Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15578834 Supplier Avantor J.T.Baker Supplier No. V37503

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High purity solvents for use in analytical and preparative separations

    For Use in Ion-Pair Chromatography, High Performance Liquid Chromatography

Specifications

CAS 5574-97-0
Packaging Wide Mouth Amber Glass Bottle
Molecular Formula C16H38NO4P
MDL Number MFCD00064526
Quantity 25 g
Specific Gravity 1L = 0.00kg
Synonym tetrabutylammonium dihydrogen phosphate, tetrabutylammonium phosphate, tetrabutylammonium dihydrogenphosphate, 1-butanaminium, n,n,n-tributyl-, phosphate 1:1, tetra-n-butylammonium phosphate, tetrabutylazanium dihydrogen phosphate, dihydrogen phosphate; tetrabutylazanium, dihydrogen phosphate; tetrabutylammonium, tetrabutyl ammonium dihydrogen phosphate, tetrabutylammonium ion dihydrogenphosphate ion
InChI Key ARRNBPCNZJXHRJ-UHFFFAOYSA-M
SMILES OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC
IUPAC Name dihydrogen phosphate;tetrabutylazanium
Molecular Weight (g/mol) 339.46
PubChem CID 2735142
Formula Weight 339.46
Grade HPLC
Chemical Name or Material Tetrabutylammonium Phosphate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.