missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium phosphate monobasic, 0.4M solution in acetonitrile, AcroSeal™

Product Code. 11985451 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11985451 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11985451 Supplier Thermo Scientific Acros Supplier No. 435991000
  • ONLINE EXCLUSIVE PRICE
    174.00 €
    168.00 € / 100mL
    Save 6.00 €
    3% Off
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5574-97-0 | C16H38NO4P | 339.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetrabutylammonium phosphate monobasic
CAS 5574-97-0, 75-05-8
Melting Point -48.0°C
Color Colorless to Yellow
Density 0.8280g/mL
Boiling Point 81.0°C to 82.0°C
Flash Point 2°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C16H38NO4P
Linear Formula (CH3(CH2)3)4NH2PO4
MDL Number MFCD00064526
Quantity 100 mL
Specific Gravity 0.828
Synonym tetrabutylammonium dihydrogen phosphate, tetrabutylammonium phosphate, tetrabutylammonium dihydrogenphosphate, 1-butanaminium, n,n,n-tributyl-, phosphate 1:1, tetra-n-butylammonium phosphate, tetrabutylazanium dihydrogen phosphate, dihydrogen phosphate; tetrabutylazanium, dihydrogen phosphate; tetrabutylammonium, tetrabutyl ammonium dihydrogen phosphate, tetrabutylammonium ion dihydrogenphosphate ion
Solubility Information Solubility in water: soluble.
InChI Key ARRNBPCNZJXHRJ-UHFFFAOYSA-M
SMILES OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC
IUPAC Name dihydrogen phosphate;tetrabutylazanium
Molecular Weight (g/mol) 339.46
PubChem CID 2735142
Viscosity 0.35 Pas (20°C)
Formula Weight 339.45
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tetrabutylammonium phosphate monobasic, 0.4M solution in acetonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.