missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium phosphate monobasic, 0.5M conc. in HPLC grade water, HPLC grade

Product Code. 11935451 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11935451 500 mL 500mL
10526412 25 mL 25mL
10512204 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11935451 Supplier Thermo Scientific Supplier No. 420175000
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5574-97-0 | C16H38NO4P | 339.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ion pair chromatography, Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 100.0°C
Chemical Name or Material Tetrabutylammonium phosphate monobasic
CAS 5574-97-0, 7732-18-5
Physical Form Solution
Quantity 500 mL
Molecular Formula C16H38NO4P
Linear Formula (CH3(CH2)3)4NH2PO4
MDL Number MFCD00064526
Synonym tetrabutylammonium dihydrogen phosphate, tetrabutylammonium phosphate, tetrabutylammonium dihydrogenphosphate, 1-butanaminium, n,n,n-tributyl-, phosphate 1:1, tetra-n-butylammonium phosphate, tetrabutylazanium dihydrogen phosphate, dihydrogen phosphate; tetrabutylazanium, dihydrogen phosphate; tetrabutylammonium, tetrabutyl ammonium dihydrogen phosphate, tetrabutylammonium ion dihydrogenphosphate ion
InChI Key ARRNBPCNZJXHRJ-UHFFFAOYSA-M
SMILES OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC
IUPAC Name dihydrogen phosphate;tetrabutylazanium
Molecular Weight (g/mol) 339.46
PubChem CID 2735142
Formula Weight 339.45
Concentration or Composition (by Analyte or Components) 0.48 to 0.52M
Packaging Glass Bottle
Specific Gravity 1.01
Density 1.0100g/mL
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.