missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Phosphate Monobasic Solution 1.0 M in H2O, TRC

Product Code. 30449216
Change view
Click to view available options
Quantity:
1 mL
5 mL
10 mL
Unit Size:
10mL
1mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30449216 10 mL 10mL
30454621 5 mL 5mL
30451522 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30449216 Supplier Toronto Research Chemicals Supplier No. TRCT31073510ML
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5574-97-0
Molecular Formula C16 H36 N . H2 P O4
Molecular Weight (g/mol) 339.45
Synonym Tetrabutylammonium Dihydrogen Phosphate, Tbap, Tetrabutylammonium Phosphate
IUPAC Name dihydrogen phosphate;tetrabutylazanium
SMILES Cccc[N+](Cccc)(Cccc)Cccc.Op(=O)(O)[O-]

Specifications

Quantity 10 mL
InChI Formula InChI=1S/C16H36N.H3O4P/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-5(2,3)4/h5-16H2,1-4H3;(H3,1,2,3,4)/q+1;/p-1
Formula Weight 339.2538
Concentration or Composition (by Analyte or Components) 1.0 M
Chemical Name or Material Tetrabutylammonium Phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.