missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium tetrafluoroborate, 98%

Product Code. 10010951 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10010951 100 g Plastic bottle 100g
10770585 5 g Glass Bottle 5g
10123201 25 g Plastic bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10010951 Supplier Thermo Scientific Acros Supplier No. 353091000
  • ONLINE EXCLUSIVE PRICE
    203.00 €
    197.00 € / 100g
    Save 6.00 €
    2% Off
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 429-42-5 | C16H36BF4N | 329.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 429-42-5
Molecular Formula C16H36BF4N
Molecular Weight (g/mol) 329.27
MDL Number MFCD00011634
InChI Key NNZZSJSQYOFZAM-UHFFFAOYSA-N
Synonym tetrabutylammonium tetrafluoroborate, tetra-n-butylammonium tetrafluoroborate, tetrabutylammonium fluoroborate, 1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-, tetrabutyl ammonium tetrafluoroborate, tetrabutylammonium ion tetrafluoroborate, 1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-1:1, tbabf4, acmc-209jry, tbabf4;
PubChem CID 67932
IUPAC Name tetrabutylazanium;tetrafluoroborate
SMILES F[B-](F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 156°C to 163°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min
Packaging Plastic bottle
Quantity 100 g
Beilstein 04, III, 293
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,1g/10 mL methanol
Formula Weight 329.28
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium tetrafluoroborate, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.