missing translation for 'onlineSavingsMsg'
Learn More

Tetrachlorobis(tetrahydrofuran)zirconium(IV), Thermo Scientific Chemicals

Product Code. 11328358
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11328358 10 g 10g
11318358 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328358 Supplier Thermo Scientific Alfa Aesar Supplier No. 039550.09
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

It is an important organic intermediate and can be used in agrochemicals, paharmaceuticals and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21959-01-3
Molecular Formula C8H16Cl4O2Zr
Molecular Weight (g/mol) 377.238
MDL Number MFCD00145367
InChI Key VDJJKYYENYIHFF-UHFFFAOYSA-J
Synonym tetrachlorobis tetrahydrofuran zirconium, zrcl4 thf 2, oxolane; tetrachlorozirconium, tetrachlorobis tetrahydrofurane zirconium, zirconium tetrachloride tetrahydrofuran complex, zirconium iv chloride tetrahydrofuran complex, bis tetrahydrofuran ; zirconium iv chloride, zirconium iv chloride tetrahydrofuran complex 1:2
PubChem CID 2734040
IUPAC Name oxolane;tetrachlorozirconium
SMILES C1CCOC1.C1CCOC1.Cl[Zr](Cl)(Cl)Cl

Specifications

Melting Point 175°C to 177°C
Color White
Quantity 10 g
Physical Form Crystalline
UN Number UN2925
Solubility Information Reacts with water.
Formula Weight 377.25
Chemical Name or Material Tetrachlorobis(tetrahydrofuran)zirconium(IV)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.