missing translation for 'onlineSavingsMsg'
Learn More

Tetrachloroethylene, Extra Pure, SLR, Fisher Chemical™

Product Code. 10234210 Shop All Fisher Chemical Products
Click to view available options
Quantity:
2.5 L
5 L
25 L
Packaging:
Amber glass bottle
Metal can
Plastic Coated Glass bottle
Unit Size:
2.5 L
25 L
5 L
This item is not returnable. View return policy

Product Code. 10234210

Brand: Fisher Chemical T/0600/21

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 127-18-4
Molecular Formula C2Cl4
Molecular Weight (g/mol) 165.82
MDL Number MFCD00000834
InChI Key CYTYCFOTNPOANT-UHFFFAOYSA-N
Synonym tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin
PubChem CID 31373
ChEBI CHEBI:17300
IUPAC Name 1,1,2,2-tetrachloroethene
SMILES ClC(Cl)=C(Cl)Cl

Specifications

Melting Point -22°C
Color Colorless
Boiling Point 120°C
Packaging Metal can
Quantity 5 L
Formula Weight 165.83g/mol
Vapor Pressure 18mbar at 20°C
Physical Form Liquid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetrachloroethylene
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H411-Toxic to aquatic life with long lasting effects.
  • H336-May cause drowsiness or dizziness.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.