missing translation for 'onlineSavingsMsg'
Learn More

Tetrachloroethylene, TRC

Product Code. 30424043
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30424043 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30424043 Supplier Toronto Research Chemicals Supplier No. TRCT29157050ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetrachloroethene
CAS 127-18-4
Molecular Formula C2 Cl4
Quantity 50 mL
Synonym Ethene','1,1,2,2-tetrachloro-, Ethene','tetrachloro- (9CI), Ethylene','tetrachloro- (8CI), 1,1,2,2-Tetrachloroethene, 1,1,2,2-Tetrachloroethylene, Ankilostin, Antisal 1, Asahi Perchlor, Didakene, Dilatin PT, Ethylene tetrachloride, F 1110, F 1110 (halocarbon), Fedal-Un, Freon 1110, LXGL 15, NSC 9777, Nema, PCE, PCE (chlorohydrocarbon), PerSec, Perchlorethylene, Perchloroethene, Perchloroethylene, Perclene, Perklone, Persa P 3, R 1110, Tetlen, Tetracap, Tetrachlorethylene, Tetrachloroethene, Tetrachloroethylene, Tetraguer, Tetraleno, Tetropil
InChI Formula InChI=1S/C2Cl4/c3-1(4)2(5)6
SMILES ClC(=C(Cl)Cl)Cl
IUPAC Name 1,1,2,2-tetrachloroethene
Molecular Weight (g/mol) 165.83
Formula Weight 163.8754
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.