missing translation for 'onlineSavingsMsg'
Learn More

Tetrachloroethylene, ULTRA RESI-ANALYZED™, J.T. Baker™

Product Code. 15107193 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
Unit Size:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15107193 4 L Narrow Mouth Amber Glass Bottle 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15107193 Supplier Avantor J.T.Baker Supplier No. 936003
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High purity solvents and salts developed for organic residue extraction/concentration procedures

    For use in Gas Chromatography (GC), Trace Hydrocarbon Analysis by IR Spectrophotometry; For Laboratory, Research, or Manufacturing Use

Specifications

CAS 127-18-4
Packaging Narrow Mouth Amber Glass Bottle
Molecular Formula C2Cl4
MDL Number MFCD00000834
Quantity 4 L
Specific Gravity 1L = 1.62kg
Synonym tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin
InChI Key CYTYCFOTNPOANT-UHFFFAOYSA-N
SMILES ClC(Cl)=C(Cl)Cl
IUPAC Name 1,1,2,2-tetrachloroethene
Molecular Weight (g/mol) 165.82
PubChem CID 31373
ChEBI CHEBI:17300
Formula Weight 165.83
Grade ULTRA RESI-ANALYZED
Chemical Name or Material Tetrachloroethylene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • tetrachloroethylene
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P391-Collect spillage.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.