Product Code. 11974401
Shop All Thermo Scientific Chemicals Products
Click to view available options
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Packaging
Unit Size
11974401
2.5 g
Glass bottle
2.5g
10155370
500 mg
Glass bottle
500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code.
11974401
Supplier
Thermo Scientific Acros
Supplier No.
309160025
193.80 EUR valid until 2026-12-31 BEST PRICE on Promo! Use promo code "27993" to get your promotional price.
missing translation for 'validMainframeMsg'
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
CAS: 4181-95-7 | C40H82 | 563.10 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
CAS
4181-95-7
Molecular Formula
C40H82
Molecular Weight (g/mol)
563.10
MDL Number
MFCD00015267
InChI Key
KUPLEGDPSCCPJI-UHFFFAOYSA-N
Synonym
n-tetracontane, unii-i79s9ixb9y, alkane c40, i79s9ixb9y, tetracontane, analytical standard, acmc-209jlw, n-tetracontane 500mg, tetracontane gc
PubChem CID
20149
IUPAC Name
tetracontane
SMILES
CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC
Specifications
Melting Point
80.0°C to 83.0°C
Color
White
Infrared Spectrum
Authentic
Assay Percent Range
97.5% min. (GC)
Packaging
Glass bottle
Linear Formula
CH3 (CH2 )38 CH3
Quantity
2.5 g
Beilstein
01,III,592
Solubility Information
Solubility in water: insoluble. Other solubilities: soluble in petroleum ether
Formula Weight
563.1
Percent Purity
98%
Physical Form
Powder or Flaky Solid
Chemical Name or Material
Tetracontane
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?