missing translation for 'onlineSavingsMsg'
Learn More

Tetracycline, TRC

Product Code. 30011958
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011958 25 g 25g
30007248 100 g 100g
30008280 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011958 Supplier Toronto Research Chemicals Supplier No. TRCT29141325G
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-54-8
Molecular Formula C22 H24 N2 O8
Molecular Weight (g/mol) 444.43
Synonym 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo- (7CI,8CI), 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-, [4S-(4α,4aα,5aα,6β,12aα)]-, (4S,4aS,5aS,6S,12aS)-4-(Dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide, (-)-Tetracycline, Abramycin, Achromycin, Achromycin (naphthacene derivative), Acromicina, Actisite, Agromicina, Akne-Pyodron, Akne-Pyodron Kur, Ambramicina, Ambramycin, Apo-Tetra, Arcanacycline, Bio-Tetra, Biocycline, Britaciclina, Calociclina, Ciclibion, Cyclomycin, Cytome, Deschlorobiomycin, Dispatetrin, Economycin, Enterocycline, Florocycline, Gammatet, Ibicyn, Kinciclina, Latycin, Limecycline, Medocycline, Mericycline, Micycline, NSC 108579, Neocycline, NorTet, Novo-Tetra, Nu-Tetra, Omegamycin, Orlycycline, Panmycin, Polycycline, Polycycline (antibiotic), Quimocyclin N, Quimpe Antibiotico, Resteclin, Retet, Robitet Robicaps, Roviciclina, Sagittacin N, Servitet, Spaciclina, Steclin V, Sumycin syrup, Tetra Hubber, Tetra-Co, Tetra-Proter, Tetrabioptal, Tetracap, Tetracitro S, Tetracycline, Tetradecin, Tetrafil, Tetrafosammina, Tetralan, Tetralen, Tetramig, Tetramykoin, Tetraseptine, Triphacycline, Veracin, Vetacyclinum
IUPAC Name (4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide
SMILES CN(C)[C@H]1[C@@H]2C[C@H]3C(=C(O)[C@]2(O)C(=O)C(=C1O)C(=O)N)C(=O)c4c(O)cccc4[C@@]3(C)O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C22H24N2O8/c1-21(31)8-5-4-6-11(25)12(8)16(26)13-9(21)7-10-15(24(2)3)17(27)14(20(23)30)19(29)22(10,32)18(13)28/h4-6,9-10,15,25,27-28,31-32H,7H2,1-3H3,(H2,23,30)/t9-,10-,15-,21+,22-/m0/s1
Formula Weight 444.1533
Chemical Name or Material Tetracycline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.