missing translation for 'onlineSavingsMsg'
Learn More

Tetradecanamine, TRC

Product Code. 30411004
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30411004 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30411004 Supplier Toronto Research Chemicals Supplier No. TRCT29153010G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetradecylamine
CAS 2016-42-4
Molecular Formula C14 H31 N
Quantity 10 g
Synonym 1-Tetradecanamine (9CI','ACI), Tetradecylamine (6CI','8CI), 1-Aminotetradecane, 1-Tetradecylamine, Alamine 5D, Armeen 14, Armeen 14D, Farmin 40D, Monotetradecylamine, Myristylamine, n-Tetradecylamine, Nissan Amine MB, NSC 66437, Tetradecanamine
InChI Formula InChI=1S/C14H31N/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15/h2-15H2,1H3
SMILES CCCCCCCCCCCCCCN
IUPAC Name tetradecan-1-amine
Molecular Weight (g/mol) 213.4
Formula Weight 213.2457
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.