missing translation for 'onlineSavingsMsg'
Learn More

Tetradecanedioic acid, 98%

Product Code. 15486728
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15486728

Brand: Thermo Scientific Alfa Aesar H60254.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 821-38-5 | C14H26O4 | 258.358 g/mol

Tetradecanedioic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetradecanedioic acid is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, reducing agents amnd bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 821-38-5
Molecular Formula C14H26O4
Molecular Weight (g/mol) 258.358
MDL Number MFCD00002743
InChI Key HQHCYKULIHKCEB-UHFFFAOYSA-N
Synonym 1,12-dodecanedicarboxylic acid, tetradecane-1,14-dioic acid, 1,14-tetradecanedioic acid, unii-uec3ac47h1, dodecamethylenedicarboxylic acid, tetradecanedicarboxylic acid, uec3ac47h1, 1,12-dodecanedicarboxylicacid, tetradecanedicarboxylate, pubchem18099
PubChem CID 13185
ChEBI CHEBI:76308
IUPAC Name tetradecanedioic acid
SMILES C(CCCCCCC(=O)O)CCCCCC(=O)O

Specifications

Melting Point 124°C to 127°C
Linear Formula HOOC(CH2)12COOH
Quantity 5 g
Beilstein 1788202
Solubility Information Slightly soluble in water.
Formula Weight 258.35
Percent Purity 98%
Chemical Name or Material Tetradecanedioic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.