missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetradecanedioic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 1,12-Dodecanedicarboxylic acid |
| CAS | 821-38-5 |
| Molecular Formula | C14 H26 O4 |
| Quantity | 250 mg |
| Synonym | Tetradecanedioic acid (6CI','7CI','8CI','9CI','ACI), 1,12-Dicarboxydodecane, 1,12-Dodecanedicarboxylic acid, 1,14-Tetradecanedioic acid, DCA 14, Emerox 114, NSC 9504 |
| InChI Formula | InChI=1S/C14H26O4/c15-13(16)11-9-7-5-3-1-2-4-6-8-10-12-14(17)18/h1-12H2,(H,15,16)(H,17,18) |
| SMILES | OC(=O)CCCCCCCCCCCCC(=O)O |
| IUPAC Name | tetradecanedioic acid |
| Molecular Weight (g/mol) | 258.35 |
| Formula Weight | 258.1831 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?