missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetradecanoic-2,2-d2 Acid, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Tetradecanoic-2,2-D2 Acid |
| CAS | 30719-21-2 |
| Molecular Formula | C14 D2 H26 O2 |
| Quantity | 0.5 g |
| Synonym | Tetradecanoic-2,2-d2 acid (9 CI), Myristic-2,2-d2 acid (8 CI), 2,2-Dideuterio-tetradecanoic acid, [2,2-2H2]-Myristic acid, α-Dideuterotetradecanoic acid, Tetradecanoic acid-2,2-d2, Tetradecanoic-2,2-D2 acid |
| InChI Formula | InChI=1 S/C14H28O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14(15)16/h2-13H2,1H3,(H,15,16)/i13D2 |
| SMILES | [2 H]C([2 H])(CCCCCCCCCCCC)C(=O)O |
| IUPAC Name | 2,2-dideuteriotetradecanoic acid |
| Molecular Weight (g/mol) | 230.383 |
| Formula Weight | 230.221 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction