missing translation for 'onlineSavingsMsg'
Learn More

Tetradecyl sodium sulfate, 95%

Product Code. 11944571 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11944571 25 g Glass bottle 25g
10458862 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11944571 Supplier Thermo Scientific Acros Supplier No. 420260250
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1191-50-0 | C14H29NaO4S | 316.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1191-50-0
Molecular Formula C14H29NaO4S
Molecular Weight (g/mol) 316.43
MDL Number MFCD00007468
InChI Key UPUIQOIQVMNQAP-UHFFFAOYSA-M
Synonym sodium tetradecyl sulfate, sodium myristyl sulfate, niaproof 4, sodium tetradecyl sulphate, tesapon k 14, texapon k 14, trombavar, natri tetradecylsulfas, sotradecol sodium, tetradecyl sodium sulfate
PubChem CID 23665770
IUPAC Name sodium;tetradecyl sulfate
SMILES [Na+].CCCCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Melting Point 199.0°C
Color White
Elemental Analysis 94% min.
Infrared Spectrum Authentic
Assay Percent Range 94% min
Packaging Glass bottle
Linear Formula CH3(CH2)13OSO3Na
Quantity 25 g
Formula Weight 316.44
Percent Purity 95%
Physical Form Micro-crystalline Powder
Chemical Name or Material Tetradecyl sodium sulfate
Show More Show Less
compliance-icons
Product Identifier
  • Tetradecyl sodium sulphate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Sodium tetradecyl sulphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.