missing translation for 'onlineSavingsMsg'
Learn More

Tetradecylbenzene, 98%, Acros Organics™

Product Code. 10564901 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Glass bottle; 5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10564901 Glass bottle; 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10564901 Supplier Thermo Scientific Acros Supplier No. 124680050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolitik

Tekniske data

CAS 1459-10-5
Melting Point 15.0°C
Color Colorless to Yellow
Density 0.8500g/mL
Boiling Point 359.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C20H34
Linear Formula C6H5(CH2)13CH3
Refractive Index 1.4805 to 1.4825
MDL Number MFCD00015080
Quantity 5 g
Specific Gravity 0.85
Synonym 1-phenyltetradecane, benzene, tetradecyl, 1-tetradecylbenzene, tetradecane, 1-phenyl, myristylbenzene, n-tetradecylbenzene, unii-i0y9j0o95a, phenyl-n-tetradecane, acmc-209cvz, 1-phenyltetradecane, analytical standard
InChI Key JZALLXAUNPOCEU-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCCC1=CC=CC=C1
IUPAC Name tetradecylbenzene
Molecular Weight (g/mol) 274.492
PubChem CID 15086
Formula Weight 274.49
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Tetradecylbenzene, 99%
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.