missing translation for 'onlineSavingsMsg'
Learn More

Tetraethyl (dimethylaminomethylene)diphosphonate, 97%

Product Code. 11431988
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11431988 25 g 25g
11421988 5 g 5g
11441988 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11431988 Supplier Thermo Scientific Alfa Aesar Supplier No. A14581.14
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 18855-52-2 | C11H27NO6P2 | 331.286 g/mol

Tetraethyl (dimethylaminomethylene)diphosphonate undergoes Horner-Wadsworth-Emmons olefination with an aldehyde gives the ?-phosphonato enamine, readily convertible to the homologous acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraethyl (dimethylaminomethylene)diphosphonate undergoes Horner-Wadsworth-Emmons olefination with an aldehyde gives the ɑ-phosphonato enamine, readily convertible to the homologous acid.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool, dry conditions in a well sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18855-52-2
Molecular Formula C11H27NO6P2
Molecular Weight (g/mol) 331.286
MDL Number MFCD00015128
InChI Key SQDDGOVNJMIFFT-UHFFFAOYSA-N
Synonym tetraethyl dimethylaminomethylenediphosphonate, tetraethyl dimethylaminomethylene diphosphonate, phosphonic acid, dimethylamino methylene bis-, tetraethyl ester, tetraethyl dimethylamino methylene bisphosphonate, acmc-1bs7f, 1,1-bis diethoxyphosphoryl-n,n-dimethylmethanamine, dimethylamino methylene bis phosphonic acid diethyl ester, diethyl diethoxyphosphoryl dimethylamino methylphosphonate, diethoxy-phosphoryl-dimethylamino-methyl-phosphonic acid diethyl ester
PubChem CID 86795
IUPAC Name 1,1-bis(diethoxyphosphoryl)-N,N-dimethylmethanamine
SMILES CCOP(=O)(C(N(C)C)P(=O)(OCC)OCC)OCC

Specifications

Boiling Point 138°C to 140°C (1 mmHg)
Refractive Index 1.451
Quantity 25 g
Beilstein 1914890
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 331.29
Percent Purity 97%
Chemical Name or Material Tetraethyl (dimethylaminomethylene)diphosphonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.