missing translation for 'onlineSavingsMsg'
Learn More

Tetraethyl Methylidenebisphosphonate, TRC

Product Code. 30415574
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30415574 5 g -
30410785 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30415574 Supplier Toronto Research Chemicals Supplier No. TRCT2928005G
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1660-94-2
Molecular Formula C9H22O6P2
Molecular Weight (g/mol) 288.21
Synonym P,P'-Methylenebisphosphonic Acid P,P,P',P'-Tetraethyl Ester, Methylenebisphosphonic Acid Tetraethyl Ester, Bis(diethylphosphono)methane, Methylenebis(diethoxyphosphine oxide), Methylenebis(diethyl phosphonate), Methylenebis(phosphonic acid) tetraethyl Ester, Tetraethyl methylenediphosphonate
IUPAC Name 1-[diethoxyphosphorylmethyl(ethoxy)phosphoryl]oxyethane
SMILES CCOP(=O)(CP(=O)(OCC)OCC)OCC

Specifications

Quantity 5 g
InChI Formula InChI=1S/C9H22O6P2/c1-5-12-16(10,13-6-2)9-17(11,14-7-3)15-8-4/h5-9H2,1-4H3
Formula Weight 288.0892
Chemical Name or Material Tetraethyl Methylidenebisphosphonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.