missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraethylammonium acetate tetrahydrate, 96%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
Specifications
Specifications
| Melting Point | 40.0°C to 45.0°C |
| Color | Colorless to White |
| Flash Point | 60°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97+% |
| Packaging | Glass bottle |
| Linear Formula | (C2H5)4N(O2CCH3)·4H2O |
| Quantity | 500 g |
| Solubility Information | Solubility in water: soluble. |
| Formula Weight | 261.35 |
| Show More |
Safety and Handling
Product Identifier
- Tetraethylammonium acetate tetrahydrate
- Danger
- Flammable solid Category 1
- H228-Flammable solid.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction