missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium bromide, 98%

Product Code. 11424934
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11424934 250 g 250g
11414934 1000 g 1000g
11434934 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11424934 Supplier Thermo Scientific Alfa Aesar Supplier No. A13835.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 71-91-0 | C8H20BrN | 210.159 g/mol

Tetraethylammonium bromide is used as a phase transfer catalyst. It acts as a source of tetraethylammonium ions in pharmacological and physiological studies. It is also employed in organic chemical synthesis. It plays a major role in the preparation of tetraethylammonium superoxide from potassium superoxide for the conversion of primary alkyl halides to dialkyl peroxides. Furthermore, It is utilized as a catalyst in the oxidation of organic sulfide to sulfoxide using o-iodoxybenzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraethylammonium bromide is used as a phase transfer catalyst. It acts as a source of tetraethylammonium ions in pharmacological and physiological studies. It is also employed in organic chemical synthesis. It plays a major role in the preparation of tetraethylammonium superoxide from potassium superoxide for the conversion of primary alkyl halides to dialkyl peroxides. Furthermore, It is utilized as a catalyst in the oxidation of organic sulfide to sulfoxide using o-iodoxybenzoic acid.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-91-0
Molecular Formula C8H20BrN
Molecular Weight (g/mol) 210.159
MDL Number MFCD00011825
InChI Key HWCKGOZZJDHMNC-UHFFFAOYSA-M
Synonym tetraethylammonium bromide, teab, tea bromide, tetrylammonium bromide, sympatektoman, tetranium, etylon, bromethyl, beparon, etambro
PubChem CID 6285
IUPAC Name tetraethylazanium;bromide
SMILES CC[N+](CC)(CC)CC.[Br-]

Specifications

Melting Point ∼285°C (decomposition)
Density 1.397
Odor Mild
Quantity 250 g
Beilstein 3563430
Sensitivity Hygroscopic
Merck Index 14,9199
Solubility Information Soluble in water.
Formula Weight 210.16
Percent Purity 98%
Chemical Name or Material Tetraethylammonium bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.