missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium chloride hydrate, 99%

Product Code. 10522551 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10522551 100 g Glass bottle 100g
10686732 25 g Glass bottle 25g
10406800 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10522551 Supplier Thermo Scientific Acros Supplier No. 150901000
  • ONLINE EXCLUSIVE PRICE
    96.10 €
    90.10 € / 100g
    Save 6.00 €
    6% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1015691-13-0 | C8H22ClNO | 183.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1015691-13-0
Molecular Formula C8H22ClNO
Molecular Weight (g/mol) 183.72
MDL Number MFCD00149992
InChI Key CALLTGJPWMIDPC-UHFFFAOYSA-M
Synonym tetraethylammonium chloride, tetraethyl ammonium chloride, etamon chloride, tea chloride, teac, n,n,n-triethylethanaminium chloride, tetraethylammoniumchloride, ethanaminium, n,n,n-triethyl-, chloride, tetraethyl-ammonium chloride, unii-8b82z83xnn
PubChem CID 5946
ChEBI CHEBI:78161
SMILES O.[Cl-].CC[N+](CC)(CC)CC

Specifications

Color White
Loss on Drying 10% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (on dry substance) (Argentometry)
Packaging Glass bottle
Linear Formula (C2H5)4NCl·xH2O
Quantity 100 g
Merck Index 15, 9345
Solubility Information Solubility in water: in water: 1410g/L (25°C). Other solubilities: freely soluble in alcohol, chloroform and acetone, slightly soluble in benzene and ether
IUPAC Name tetraethylazanium;chloride
Formula Weight 165.71
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Tetraethylammonium chloride hydrate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Tetraethylammonium chloride hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.