missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium fluoride hydrate, 98%

Product Code. 10388730 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10388730 25 g Glass bottle 25g
10340544 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10388730 Supplier Thermo Scientific Acros Supplier No. 216660250
  • ONLINE EXCLUSIVE PRICE
    122.00 €
    118.00 € / 25g
    Save 4.00 €
    3% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 98330-04-2 | C8H20FN | 149.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98330-04-2
Molecular Formula C8H20FN
Molecular Weight (g/mol) 149.25
MDL Number MFCD00149990
InChI Key QSUJAUYJBJRLKV-UHFFFAOYSA-M
Synonym tetraethylammonium fluoride hydrate, tetraethylammonium hydrate fluoride, tetraethylammoniumfluoridehydrate, acmc-209nvn, et4nf.h2o, tetraethylazanium fluoride hydrate, tetraethyl ammonium fluoride hydrate, ethanaminium, n,n,n-triethyl-, fluoride, monohydrate
PubChem CID 16211652
SMILES [F-].CC[N+](CC)(CC)CC

Specifications

Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (C2H5)4NF·xH2O
Quantity 25 g
IUPAC Name tetraethylazanium fluoride
Formula Weight 149.14
Percent Purity 98%
Physical Form Adhering Crystals, Crystals or Crystalline Solid
Chemical Name or Material Tetraethylammonium fluoride hydrate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Tetraethylammonium fluoride hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.