missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium hydrogensulfate, 99+%, HPLC grade

Product Code. 10519463 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10519463 50 g Glass bottle 50g
10112961 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10519463 Supplier Thermo Scientific Supplier No. 343820500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 16873-13-5 | C8H21NO4S | 227.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ion pair chromatography, Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16873-13-5
Molecular Formula C8H21NO4S
Molecular Weight (g/mol) 227.32
MDL Number MFCD00036150
InChI Key CREVBWLEPKAZBH-UHFFFAOYSA-M
Synonym tetraethylammonium hydrogensulfate, tetraethylammonium hydrogen sulphate, tetraethylammonium hydrogen sulfate, n,n,n-triethylethanaminium hydrogen sulfate 1:1:1, hydrogen sulfate; tetraethylazanium, tertraethylammonium hydrogen sulfate, tetraethyl ammonium hydrogen sulphate
PubChem CID 167582
IUPAC Name hydrogen sulfate;tetraethylazanium
SMILES OS([O-])(=O)=O.CC[N+](CC)(CC)CC

Specifications

Absorbance 0.02 max. (0.4 M in HPLC water) at 250nm, 0.04 max. (0.4 M in HPLC water) at 230nm, 0.04 max. (0.4 M in HPLC water) at 240nm, 0.05 max. (0.4 M in HPLC water) at 220nm, 0.1 max. (0.4 M in HPLC water) at 210nm
Melting Point 243°C to 245°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 50 g
Formula Weight 227.32
Percent Purity 99+%
Grade HPLC
Physical Form Powder
Chemical Name or Material Tetraethylammonium hydrogensulfate
Show More Show Less
compliance-icons
Product Identifier
  • Tetraethylammonium hydrogensulfate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.